TraceGOLD TG-35MS GC Columns, Thermo Scientific
Supplier: Thermo Scientific
Certificates
About this item
These GC columns have a higher phenyl content for useful elution order and retention time changes.
- Mid-polarity phase, 35% diphenyl/65% dimethyl polysiloxane
- Equivalent to USP G42 phase
Ideal for organochlorine pesticides and herbicides, pharmaceuticals, sterols, phthalate esters etc.
Customers Who Bought This Also Bought