About this item
Versatile Tygon® A-60-F temperature-resistant food and beverage tubing provides long life for everyday use and consistently outperforms general-purpose rubber tubing in a wide variety of applications
Tygon® A-60-F temperature-resistant food and beverage tubing offers long flex life in peristaltic pumps, and is temperature resistant from –75 to +275 °F. It is compatible with virtually all common sanitizers and cleaners, ozone and UV light resistant, repeatedly autoclavable, and chemically compatible with a wide range of fluids.
It will not weaken or crack following years of exposure to heat and ozone. This tubing is extremely abrasion-resistant and bends easily to simplify custom component assembly. Tygon® Food Process Tubing is ideal for use in clean-in-place and steam-in-place cleaning and sterilization systems.
It is compatible with virtually all commercial cleaners and sanitizers, and can be repeatedly autoclaved up to five times without affecting the overall service life. Tygon® A-60-F temperature-resistant food and beverage tubing provides superior resistance to many corrosive fluids when compared to general-purpose rubber tubings. It offers excellent acid and alkali resistance and is compatible with numerous oxidizing agents such as peroxide, hypochlorite, and ozone.
This tubing also exhibits good resistance to the animal and vegetable oils frequently found in food contact applications.
Specifications
- Catalog No:
- 63008-824
- 63008-820
- Model:
- 63008-824
- 63008-820
- Durometer hardness:
- Shore A, 61
- Shore A, 61
- Max. vacuum:
- 26.0"
- 11.0"
- Working pressure:
- 10.0 psi
- 7.0 psi