Trays, Round, Polypropylene, Dynalon
Catalog #: 13450-770
Supplier: Dynalon
Specifications
- Cat. no.:13450-770
- Interior Dimensions:10"W x 3"H
- Supplier No.:109844
Specifications
About this item
Fabricated trays of Polypropylene are heavy wall (3/16"), corrosion resistant and may be autoclaved
Trays can be customized with spigots and covers which are fabricated to order
Ordering information: Additional sizes and materials are available upon request.
Customers Who Bought This Also Bought