Trays, Dispensing, High Density Polyethylene, Dynalon
Supplier: Dynalon
Certificates
About this item
Heavy trays fabricated with 1/4" thick high density polyethylene
All welded construction. Dispensing trays are supplied with polypropylene dispensing valve located as close to the tray bottom as possible (can be in other locations upon request). All trays can be customized with spigots and covers which are fabricated to order
Ordering information: Additional sizes and materials are available upon request.
Customers Who Bought This Also Bought