Order Entry
ContactUsLinkComponent
Soil Standard, Volatiles in Soil, ERA (Waters)
Soil Standard, Volatiles in Soil, ERA (Waters)
Catalog #: CA11022-910
Supplier:  ERA
CAS Number:  
Soil Standard, Volatiles in Soil, ERA (Waters)
Catalog #: CA11022-910
Supplier:  ERA
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Certified Reference Material
  • Size:
    2 ml
  • Cat. no.:
    CA11022-910
  • Supplier No.:
    721
  • Parameters:
    Volatiles in Soil

Specifications

About this item

One 2 mL flame-sealed ampule on this volatiles in soil, soil standard in methanol requires spiking into the provided ten grams of solid matrix before analysis

Acetone, Acetonitrile, Acrolein, Benzene, Bromobenzene, Bromochloromethane, Bromodichloromethane, Bromoform, Bromomethane, 2-Butanone (MEK), n-Butylbenzene, sec-Butylbenzene, tert-Butylbenzene, Carbon disulfide, Carbon tetrachloride, Chlorobenzene, Chlorodibromomethane, Chloroethane, 2-Chloroethyl vinyl ether, Chloroform, Chloromethane, 2-chlorotoluene, 4-chlorotoluene, 1,2-Dibromo-3-chloropropane, (DBCP), 1,2-Dibromoethane (EDB), Dibromomethane, 1,2-Dichlorobenzene, 1,3-Dichlorobenzene, 1,4-Dichlorobenzene, Dichlorodifluoromethane, 1,1-Dichloroethane, 1,2-Dichloroethane, 1,1-Dichloroethylene, cis-1,2-Dichloroethylene, trans-1,2-Dichloroethylene, 1,2-Dichloropropane, 1,3-Dichloropropane, 2,2-Dichloropropane, 1,1-Dichloropropene, cis-1,3-Dichloropropylene, trans-1,3-Dichloropropylene, Ethylbenzenel, Hexachlorobutadiene, Hexachloroethane, 2-Hexanone, Isopropylbenzene, p-Isopropyltoluene, Methyl tert-butyl ether (MTBE), 4-Methyl-2-pentanone (MIBK), Methylene chloride, Naphthalene, Nitrobenzene, n-Propylbenzene, Styrene, 1,1,1,2-Tetrachloroethane, 1,1,2,2-Tetrachloroethane, Tetrachloroethene, Toluene, 1,2,3-Trichlorobenzene, 1,2,4-Trichlorobenzene, 1,1,1-Trichloroethane, 1,1,2-Trichloroethane, Trichloroethene, Trichlorofluoromethane, 1,2,3-Trichloropropane, 1,2,4-Trimethylbenzene, 1,3,5-Trimethylbenzene, Vinyl acetate, Vinyl chloride, m&p-Xylene, o-Xylene, Xylenes total.

Use with EPA methods 8021 and 8260. This standard includes a subset of analytes listed below at 20 to 200 µg/kg. Total xylenes is listed at 40 to 400 µg/kg, selected ketones are listed at 80 to 1,000 µg/kg, and acetonitrile is listed at 200 to 1,000 µg/kg.

Ordering information: This standard is not compliant with the NELAC concentration for hexachloroethane, hexachlorobutadiene or nitrobenzene.
Customers Who Bought This Also Bought