SP Bel-Art Glove Box Clamping Rings, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
Adjustable rings hold gloves in place.
- Fully adjustable rings
- Fit glove boxes with a range of port hole sizes
- Sold individually or in pairs depending on model
47735-510 fit any glove box with 6" diameter glove port holes.
47748-192 fit any glove box with 8" diameter glove port holes. Sold per each.
Ordering information: Sold per pair.
Customers Who Bought This Also Bought