To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Certified Reference Material
- Size:2 ml
- Cat. no.:CA11022-846
- Supplier No.:703
- Parameters:Regulated Volatiles
Specifications
About this item
One 2 mL flame-sealed ampule of this water supply, regulated volatiles standard yields in excess of 200 mL after dilution
Benzene, Carbon tetrachloride, Chlorobenzene, 1,2-Dichlorobenzene, 1,4-Dichlorobenzene, 1,2-Dichloroethane, 1,1-Dichloroethylene, cis-1,2-Dichloroethylene, trans-1,2-Dichloroethylene, 1,2-Dichloropropane, Ethylbenzene, Methylene chloride, Styrene, Tetrachloroethylene, Toluene, 1,2,4-Trichlorobenzene, 1,1,1-Trichloroethane, 1,1,2-Trichloroethane, Trichloroethylene, Vinyl chloride, Xylenes total
Use with EPA methods 502.2 and 524.2. This standard contains all of the analytes listed below at 2 to 50 µg/L.