Order Entry
ContactUsLinkComponent
Water Pollution Standard, PAHs – GC/GCMS, ERA (Waters)
Water Pollution Standard, PAHs – GC/GCMS, ERA (Waters)
Catalog #: CA11022-796
Supplier:  ERA
CAS Number:  
Water Pollution Standard, PAHs – GC/GCMS, ERA (Waters)
Catalog #: CA11022-796
Supplier:  ERA
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Certified Reference Material
  • Size:
    2 ml
  • Cat. no.:
    CA11022-796
  • Supplier No.:
    4882
  • Parameters:
    PAHs – GC/GCMS

Specifications

About this item

One 2 mL flame-sealed ampule yields up to 2 L of this water pollution, PAHs - GC/GCMS standard after dilution

Acenaphthene, Acenaphthylene, Anthracene, Benzo(a)anthracene, Benzo(a)pyrene, Benzo(b)fluoranthene, Benzo(k)fluoranthene, Benzo(g,h,i)perylene, Chrysene, Dibenz(a,h)anthracene, Fluoranthene, Fluorene, Indeno(1,2,3-cd)pyrene, Naphthalene, Phenanthrene, Pyrene.

Use with EPA methods 625, 8100 and 8270. Each standard contains a subset of the analytes listed below at 10 to 200 µg/L.

Customers Who Bought This Also Bought