Soil Standard, Hexavalent Chromium in Soil, ERA (Waters)
Catalog #: CA11022-714
Supplier: ERA
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Certified Reference Material
- Size:40 g
- Hexavalent chromium:40-300 mg/kg
- Cat. no.:CA11022-714
- Supplier No.:921
- Parameters:Hexavalent Chromium in Soil
Specifications
About this item
One 40 g hexavalent chromium in soil, soil standard in a screw-cap bottle can be used for all promulgated hexavalent chromium methods.
Customers Who Bought This Also Bought