About this item
This antibody reacts with human factor VIII related antigen. It stains endothelial cells and megakaryocytes in various human tissues. This antibody does not cross-react with any other cell types.
Type:
Antigen:
Clonality:
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA99991-068
- CA95025-012
- CA99991-220
- Antigen symbol:
- Factor VIII R Antigen
- Factor VIII R Antigen
- Factor VIII R Antigen
- Antigen name:
- Coagulation Factor 8 Related
- Coagulation Factor 8 Related
- Coagulation Factor 8 Related
- Clonality:
- Polyclonal
- Polyclonal
- Polyclonal
- Host:
- Rabbit
- Rabbit
- Rabbit
- Concentration:
- Ready to use
- Concentrated
- Concentrated