About this item
This antibody reacts with a protein of 81 kDa. Ezrin serves as a tyrosine kinase substrate and is phosphorylated in EGF-stimulated cells. Ezrin is a cytoplasmic protein, enriched in microvilli and other cell surfaces. Ezrin has an actin-binding capacity. Ezrin is expressed in epithelial cells but not in mesenchymal cells. Ezrin is also expressed by certain epithelial tumors, such as renal cell adenocarcinomas.
Type:
Antigen: EZR
Clonality: Monoclonal
Clone: 3C12
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity:
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- CA95024-548
- CA99990-708
- Antigen symbol:
- EZR
- EZR
- Antigen name:
- Ezrin
- Ezrin
- Antigen synonyms:
- HEL-S-105|Cytovillin|VIL2|CVIL|CVL
- HEL-S-105|Cytovillin|VIL2|CVIL|CVL
- Clonality:
- Monoclonal
- Monoclonal
- Clone:
- 3C12
- 3C12
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1
- IgG1