Specifications
- Description:Dial Vernier Caliper Metric Scale
- Supplier No.:62065-B
Specifications
About this item
B Dial type: 0.1 mm dial, 33 mm (1.3") diameter, maximum reading of 150 mm, and 10 mm bar scale.
These precision vernier calipers are molded in corrosion resistant fiberglass-filled super-polyamide nylon alloy. They are excellent tools due to the easy black numbers on white background reading scale and light weight, fine adjustment thumb roller dials. Its parallel jaws are 40 mm (1.57") deep and its reading scale does not create a parallax. Its inside-outside jaws and round metal rod depth gauge add to its value. Overall length: 213 mm (8 3/4").