About this item
The specimen basket is made of brass with a stainless steel mesh attached to both ends.
The mesh opening is 300 µm. To use, simply screw both halves together and you can keep small specimens for long periods of time during the process of fixing, dehydration or even critical point drying. It can also be used to hold small parts for ultrasonic cleaning.
Inside diameter: 12.5 mm, height: 15 mm
Outside diameter:16 mm, height: 17 mm