ContactUsLinkComponent
Vernier Caliper, Electron Microscopy Sciences
Vernier Caliper, Electron Microscopy Sciences
Catalog #: 100502-462
Minority or Woman-Owned Business Enterprise
INSTANT READOUT DIGITAL CALIPER
Vernier Caliper, Electron Microscopy Sciences
Catalog #: 100502-462
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Instant Readout Digital Caliper
  • Supplier No.:
    62065-40
Specifications
About this item
This caliper is made from stainless steel and measures inside and outside in inches and millimeters.
Read-out from 0 to 6 inches with 0.001" resolution, or 0 to 150 mm with 0.01 mm resolution. Includes a zero set button, depth measuring rod and a fine adjustment thumb wheel.
Delivery: Protective carrying case and battery are included.
Customers Who Bought This Also Bought