To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Capacity:500 ml
- Ø:33 mm
- Pore size:0.22 µm
- Color:Orange
- Supplier No.:67051-52
Specifications
About this item
Choice of membrane media: Cellulose acetate (CA), for low protein binding.
- Membranes come in 0.20, 0.22 or 0.45 µm pore sizes
- Ideal for filtration of tissue culture media, biological fluids, fixation buffers and more (not for parenteral use)
Available in 33 and 45 mm neck sizes. Connector neck with unique polyethylene gasket ensures a positive seal between filtration unit and collecting bottle. Funnel has tubing adapter that fits most vacuum hoses. Units come individually packaged and are pre-sterilized by gamma irradiation, certified non-pyrogenic.
Choice of membrane media: Cellulose acetate (CA), for low protein binding. Nylon (NYL), for low extractable. Cellulose Nitrate (CN), for general purpose filtration. Or polyethersulfone (PES), for fast flow rates and low protein binding.
All four-type membranes are detergent-free, tissue culture compatible, and heat-sealed to the support grid to maximize flow rate, reduce foaming and protein denaturization.