About this item
Unique Cone-Shaped Storage Racks For Easy Sample Viewing. Easily view, organize and track up to 46 tubes including 24 - 0.5 to 2.0 ml tubes, 14 tubes up to 8ml in size including cryovials and 12x75mm culture/blood collection tubes, 7 - 15 ml tubes and 1 - 50 ml tube. Constructed of durable polypropylene and autoclavable. Use the Stacking Adapter to double available bench space. Turn the rack over and fill with water for freezing. A non-slip rubber bottom (included with each NovaChamber) keeps the water/ice contained for a portable ice chamber rack. For easy sample tracking each rack rack has molded numbers.