ContactUsLinkComponent
Cassette Sampling Stand, Electron Microscopy Sciences
Cassette Sampling Stand, Electron Microscopy Sciences
Catalog #: 102681-136
Cassette Sample Stand Abs-1
Cassette Sampling Stand, Electron Microscopy Sciences
Catalog #: 102681-136
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    Cassette Sampling Stand, ABS-1
  • Cat. no.:
    102681-136
  • Supplier No.:
    65232-ABS
Specifications
About this item
Sampling stand used to support sampling cassettes at elevated levels
  • ZA0042 Economy Sampling Stand: very nice stand, made from black anodized aluminum
  • ABS-1 Deluxe Sampling Stand: very similar to the economy stand but made in the USA from aluminum. Better durability and stability than the economy one
Two models are available to choose from:
Customers Who Bought This Also Bought