Specifications
- Description:Single place sliding guide
- L×W:7¼×4×2"
- Cat. no.:102096-958
- Supplier No.:70494-30
Specifications
About this item
Use the Pipetter Guide (70494-30 shown) with a 96- Well Color Orienter (sold separately) for added pipetting ease. Easy microplate viewing for pipetting and liquid transfer.
- Easily align a pipetter with microplate wells using our sliding guide
- Reduces eyestrain
- Compatible with multichannel or single-channel pipetters
The Sliding Guide is clearly marked with row letters for fast and easy orientation. Adjust the guide height to accommodate flat or v-bottom microplatea or PCR plates.