Disposable Bibulous Paper, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Recommended for drying microscope slides and other small lab glass items.
- Highly absorbent white fiber paper
- Lint and dust free
- Size: 4×6
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.