Hitachi S-450 SEM Mounts, Electron Microscopy Sciences
Supplier: Avantor
Certificates
About this item
M4 Tapered hole, female thread.
Head 15 mm (5/8"), Pin height: 6 mm (1/4").
Ordering information: Comes plain or with 3 divisions.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.