Specifications
- Description:Quasar Lighting System, Kit
- Supplier No.:68107-70
Specifications
About this item
QUASAR™ LS Kit includes lighting system frame, retro fit attachment pins, longer screw for OptiLOUPE™ fitted visors, 2 ʽAA’ batteries, and velcro strap.
- L.E.D. Lighting System for Optivision Head Band Magnifiers
- Custom fit lighting attachment for magnifying visions
- Patent pending design utilizes the latest in LED technology
- Reduces shadows and increases visibility in any environment
- Lights up holes and deep cavities
- Super long batteries life –light remains bright after 16 hours of straight use
- Lasts up to 96 hours solid on a single set of batteries
- Over 100,000 hour bulb life expectanct
QUASAR™ LS Kit includes lighting system frame, retro fit attachment pins, longer screw for OptiLOUPE™ fitted visors, 2 ʽAA’ batteries, and velcro strap.