Specifications
- Description:Carbide Cleaver Tool
- Overall Length:120 mm
- Supplier No.:62165-CC
Specifications
About this item
These precision hand held tools can be used for scoring and cleaving all types of optical fibers.
Carbide cleaver tool made from a solid carbide rod. It has two angled sides which intersects into a very sharp cutting edge of about 4 mm (0.160") in length. The third angle is lapped on the very tip to prevent breakage. The carbide shank is mounted into a non-rolling hexagon shaped anodized aluminum holder.The carbide part has a 2.0 mm diameter and 10.0 mm long.
Retractable diamond cleaver tool, a ground and polished natural diamond which has a 90° included angle with a relief angle of 30°. The cleaver is retained in a quality metal pen like casing with a pocket clasp. The diamond part has a 1.5 mm width and length of 3.0 mm.
Retractable sapphire cleaver tool, ground and polished natural sapphire wedge with a 60° included angle. The cleaver is retained in a quality metal pen like casing with pocket clasp. The sapphire part has a 2.0 mm width and length of 3.0 mm.