ContactUsLinkComponent
Zerostat Anti-Static Gun, Electron Microscopy Sciences
Zerostat Anti-Static Gun, Electron Microscopy Sciences
Catalog #: 100496-120
Minority or Woman-Owned Business Enterprise
ZERO-STAT ANTI-STATIC GUN
Zerostat Anti-Static Gun, Electron Microscopy Sciences
Catalog #: 100496-120
Minority or Woman-Owned Business Enterprise
Specifications
  • Description:
    Zero-Stat Anti-Static Gun
  • Supplier No.:
    60610
Specifications
About this item
The Zerostat Anti-Static Gun operates on a piezoelectric effect to emit positive and negative charges to neutralize static on any surface. This anti-static gun emits no radioactivity and does not require electricity or batteries.
Measures: 4×6"
Customers Who Bought This Also Bought