About this item
Histology diamond knives for cryo temperatures and may be used on all ultramicrotomes and microtomes with a retraction of the specimen in the return phase.
For Room Temperature:
Knives are available with a 45° angle
Section thickness range 0.2 - 5µm
Cutting edge length 4, 6, and 8mm
- Perfect sections, free of scores or compression
- Serial sections without knife change
- Thinner sections
- No knife making procedure
Histo knives are made of natural diamonds of highest quality. They guarantee the best quality and durability of the cutting edges. Knives are available in 45° angle. Section thickness range 0.2 to 5 µm. Cutting edge length 4 and 6 mm.
The histo-cryo knives are delivered in boats (for wet cryo sectioning) and triangular holders (for dry cryo sectioning), made of a special copper/nickel alloy, which guarantees the best heat/cold transmission.
For Cryo Temperatures:
Knives are available in 45° angle
Section thickness range 0.2 - 5 µm
Cutting edge length 4 and 6mm
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.