Optivison™ Head Band Magnifiers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
The Optivison™ Head Band Magnifiers are well made, lightweight and durable, and fit over eyeglasses.
- Dial adjustment headband that fits any head size
- Adjustable pivot assembly on visor is set to desired tension, so it stays in place when tilted
- Lenses are ground and polished prismatic
- Lens plate replacement is available
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.