Specifications
- Description:Glas scribe pen
- Cat. no.:100491-936
- Supplier No.:72151
Specifications
About this item
The Glascribe® Pen, with retractable tungsten carbide tip, lets you inscribe on glass, ceramics, and plastic
The Glascribe® Pen, with retractable tungsten carbide tip, lets you inscribe on glass, ceramics, and plastic. The pen has a pocket clip, so it is always handy when needed.
The pen has a pocket clip, so it is always handy when needed.