About this item
Design minimize friction, promotes reagent mixing, and facilitates uniform hybridization.
SecureSeal™ hybridization chambers provide optimum surface to volume fluid dynamics. SecureSeal™adhesive chambers form peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. RNase free, gasketed chambers provide optimum surface-to-volume fluid dynamics for hybridization assays on large or multiple specimens and genomic arrays on glass slides or film coated slides. All Hybridization Chambers have 1.5 mm diameter Ports and come with 120 Port Seals.
- Sealable access ports (1.5 mm) in the chamber surface allow for the addition and removal of reactants
- Adhesive seal tabs create leak-proof chambers that are temperature resistant
- SecureSeal™adhesive bonds chambers to glass in seconds and removes cleanly and easily even after heating
- SecureSeals™are ideally suited for protocols utilizing autoradiographic, fluorescent or cheminescent end points
- Polycarbonate HybriSlip used, 0.25 mm thick
SecureSeal™ adhesive chambers form peel-and-stick enclosures to isolate specimens affixed to glass microscope slides. RNase free, gasketed chambers provide optimum surface-to-volume fluid dynamics for hybridization assays on large or multiple specimens and genomic arrays on glass slides or film coated slides.