Polysine™ Microscope Adhesion Slide, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
The permanent bio-adhesive material on our Polysine™ slide attracts frozen and paraffin embedded sections, centrifuge preparation and cytology smears.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.