About this item
GOLDSEAL® slides are made from pure white glass (sodalime) and at accurately cut sizes.
To preserve the highest quality from manufacturer to ultimate user, all GOLD SEAL® slides are hermetically sealed in our Moisture-Barrier™ system, providing a dry-slide environments during shipment and storage.