Specifications
- Description:Extended reach micro ruler only
- Supplier No.:62136-ER
Specifications
About this item
Micro Mirror - Extended Reach
Micro Ruler: Extended Reach
Precision etched, non-glare ruler made from surgical stainless steel, only 3 mm wide. The ruler is calibrated in 0.1 mm graduations, 10 mm total range. Stainless steel handle 230 mm long. Balanced and angled for the ultimate in flexibility able to reach into areas which are impossible for other measuring devices. Available with or without a stainless steel autoclave case. Total weight of the ruler is 26 g. Applications include microscopy, chemistry and anywhere precise measurement in clean and very confined areas are of critical importance.
Micro Mirror - Extended Reach
It is similar construction with Micro-Ruler, but comes with the 2.5x magnify Mirror - made from polished stainless steel, 4.8 mm diameter. Overall length is 230 mm. Autoclavable Case is available as an option.