Thin Sample Holder, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Ideal for examination cross section of thin samples, such as wafers, multi-layer of capacitors, plastics, metals and more.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.