About this item
Unflexible grids, made of a special beryllium-copper alloy, nickel and molybdenum.
- Extreme stability for coating with support film
- Contamination free and reusable after cleaning
- Diameter 3.05 mm
- Very reliable under the electron beam
NUM grids: Numbered grids are in random order. Numbers may be duplicated.
DOT grids: 2 dots are marked on one side of the grid for identification. Dots are visible to the naked eye.
NOTCH grids: A mark, stamped on one side of the grid to facilitate the handling of the grids. Notch is available with NUM or DOT grids.
GILDED grids: Completely gold-plated grids, suitable for immunology research, autoradiography, as well as special needs.