Bottles, Square, Wide Mouth, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
These bottles are made from high density polyethylene.
They are leak-proof for both solids and liquids and are very space efficient.
Delivery: They are supplied with push-in seals and easy-to-grip screw caps which have a toggle for tamper evident wire attachment. Supplied pre-capped in shrink-wrap.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.