Specimen Discs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Accurately machined chrome plated aluminum discs that fit most microtomes and cryostats
They have a knurled surface for better specimen handling. Straight shank. Available in a variety of disc sizes.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.