Specifications
- Description:Opaque White Pen
- Cat. no.:102097-878
- Supplier No.:72168-01
Specifications
About this item
Permanently mark on nearly any surface in white color and the marking is waterproof
This pen is ideal for SEM, marking sample identification on carbon tabs, aluminum stubs, conductive tape or any similar surface. The pen contains xylene.