Specifications
- Magnification:50×
- Description:Pocket Measuring Microscope
- Supplier No.:68131
Specifications
About this item
As small as a fountain pen, the pocket measuring microscope has a standard scale is incorporated in its lens barrel, giving an effecient brightness and excellent definition. It can be focused in the same way as a conventional microscope.
- 25x magnification has 3.30 mm field of view, 3 mm measuring range and minimal scale division of 0.005
- 50x magnification has 2.00 mm field of view, 1.6 mm measuring range and minimal scale division of 0.02
Dimensions: 12.40 x 127 mm