Specifications
- Description:Micro Tube Beta Shield
- Supplier No.:64450-00
Specifications
About this item
It is ideal for DNA sequencing.
- Measures 2¹⁄₆×4⅛×2⁷⁄₁₆" (52×105×62 mm)
- Tubes not included
This shield is a solid, molded, clear-acrylic block with 13 wells 11 mm in diameter. It holds 13 of 1.5 ml micro sample tubes (EMS Cat. #72363) or similar. A clear tight-fitting cover allows space for the micro tube flip top.