Specifications
- Magnification:8×
- Description:Measuring Magnifier Mag6
- Supplier No.:68055
Specifications
About this item
A hand-held magnifier for measurement and inspection
Their construction make them ideal aids for anyone concerned with quality control, inspection, drafting, photography, etc. The clear base allows ample surface light to fall on the subject being measured and the eye lens can be focused to suit individual sight. Interchangeable graticules fitted in the base ring permit a variety of measurements to be made quickly and accurately. Comes complete with a leather case.