To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Dimensions:12×12×27" (W×L×H)
- Description:Glass Disposable Box, Floor
- Cat. no.:102091-100
- Weight:15.4 lbs.
- Supplier No.:64412
Specifications
About this item
These boxes are used to dispose of empty ampoules, broken glass, empty vials, biohazard wastes, etc.
These boxes are used to dispose of empty ampoules, broken glass, empty vials, biohazard wastes, etc. The outer box is made from corrugated cardboard; the inner one is a 2-mil polypropylene bag. When the box is full, the safety cap is pulled into place and the entire unit is ready for disposal or incineration. There are no replacement bags for these boxes.
- Colorful graphics for quick identification
- Assembly instructions on each box
- Folding design for shipping and storage
The outer box is made from corrugated cardboard, the inner one is a 2-mil polypropylene bag. When the box is full, the safety cap is pulled into place and the entire unit is ready for disposal or incineration. There are no replacement bags for these boxes.