Specifications
- Volume:284 g (10 oz.)
- Description:Dust-Off Xl(DPSXLA)
- Supplier No.:70838
Specifications
About this item
Pressurized duster removes dust and dirt with pure moisture free blasts. There is more product at less cost per ounce. Extension tube is included with the can. Its new concentrated formula will give you 57% more blasts per ounce, 17% more blasts per can. Safe for the environment. Contains: Formula S (Difluoroethane) - 100% safe for the ozone.