Specifications
- Description:Duplex Reference Specimen
- Supplier No.:79521-01
Specifications
About this item
An alternative and very sensitive test is by means of an alloy with two major Copper/Zinc phases separated by an atomic number difference of 0.1. The light phase illustrated in the micrograph has a mean atomic number of 29.47 and the dark phase a mean atomic number of 29.37.