Carbon Film on Hexagonal Grids, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
A thin film of pure carbon is deposited on the shiny side of the grid.
- Carbon coatings thickness of standard (10 nm), ultra-thin (5 to 6 nm), thick (15 to 20 nm), or extra thick (25 to 50 nm)
- Optically checked followed by batch testing in the EM
Packaging: Packed in grid storage box.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.