Specifications
- Description:Grinder/Polisher
- Supplier No.:50120-10
Specifications
About this item
This light weight, bench top Grinding/Polishing model is ideal for surface preparation of small metallographic specimens. It is adjustable in speed from 0 to 1725 RPM, and has 1/3 HP and it is fully grounded.
The coolant is applied to the wheel through an adjustable flow control goose-neck spout. The unit comes complete with a removable rubber splash guard for cleaning and a compression fitting for an easy connection into the existing coolant supply.
Measures: 13” x 15.25 x 9.5” (H); Weight 35 lb.
110 volt, 60 Hz.