About this item
This special soft coating provides a non-slip grip and improves the handling of the tweezer. The soft-coating prevents hand fatigue, and protects the hand from heat and cold. Made from the finest stainless steel grade.
Style SS measures 135 mm (5 1/3”) Long. M = 8 mm (1/3”) N = 1.5 mm (1/16”) P = 82 mm (3 ¼”). Style 1 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”) P = 50 mm (2”). Style 3 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”) P = 50 mm (2”). Style 3C measures 110 mm (4 1/3”) Long. M = 9 mm (1/3”) N = 1.5 mm (1/16”) P = 43 mm (1 2/3”). Style O measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”) P = 51 mm (2”). Style OO measures 115 mm (4 ½”) Long. M = 10 mm (1/3”) N = 2.5 mm (1/10”) P = 46 mm (1 ¾”) U = 0.5 mm (½0”). Style 2A measures 120 mm ( ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”) P = 35 mm (1 3/8”) U = 2 mm (1/12”). Style 5 measures 120 mm (4 ¾”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”) P = 30 mm (1 1/16”). Style 5A measures 115 mm (4 ½”) Long. M = 10 mm (1/3”) N = 1.5 mm (1/16”) P = 34 mm (1 1/3”). Style 7 measures 120 mm (4 ¾”) Long. M = 11 mm (½”) N = 1.5 mm (1/16”) P = 34 mm (1 1/3”).