About this item
Dumoxel is an alloy composed of C, Cr, Ni, Mo and Cu. This alloy was developed and patented by Dumont to offer the best in resistance to corrosion thanks to its high concentration of molybdenum and chromium.
- Dumoxel is 95% antimagnetic
- Resistant against temperatures of around 400 °C (DIN 50 914)
- Suitable for autoclave sterilization at 270 °C
Dumoxel also guarantees an excellent resistance to sulphuric environments, hydrochloric acid, as well as to all other mineral and organic acids.
Length: 135 mm.