Pliers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Glass-filled delrin pliers with flat smooth jaws.
- Hold tubes securely and resistant to most solvents
- Suitable for handling soft materials
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.