SafetySet, S.C.A.T.
Supplier: S.C.A.T.
Certificates
About this item
Solvent Safety Cap set suitable for use with all current HPLC systems
Set comes with four safety caps and all required connectors. Unused connectors can be closed by the blind screw delivered with the set. This makes the system universally applicable, even when less connectors are needed for a time.
Related Information
190206
Images 360
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
Customers Who Bought This Also Bought