About this item
For safe disposal of fluid waste.
Collect your solvent waste safely, without being exposed to hazardous gases and solvent vapours. The integrated exhaust filter absorbs 99% of solvent vapors, thereby protecting staff and the working environment. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
- Especially recommended for HPLC Systems
- Chemically resistant to aggressive organic solvents
Collect your solvent waste safely, without being exposed to hazardous gases and solvent vapours. The integrated exhaust filter absorbs 99% of solvent vapors, thereby protecting staff and the working environment. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
Available for multiple screw threads and container sizes.
With safety funnel: Safety Funnels are solely opened when disposing waste fluids. When releasing the opening mechanism, the waste container closes automatically. Furthermore you can discharge fluids permanently through the tubing ports (e.g. for HPLC applications).
Related Information
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
SCTV107925