Fire Resistant SafetyCaps, S.C.A.T.
Supplier: S.C.A.T.
Certificates
About this item
Fire resistant SCAT caps incorporate a self-extinguishing component with a special plastic.
- Integrated air valve with PTFE membrane for pressure equalisation while extracting fluids
- Reliable protection for operator and environment
- Air valve blocks escaping vapours and absorbs dust and dirt particles of inflowing air
In the event of a fire, the SCAT fire resistant material builds an insulating coating that acts as a heating brake. A barrier is formed which prevents oxygen from entering and so stops the flames spreading.
SafetyCaps with shut-off: With stopcock to shut-off the tubing connections when exchanging the solvent bottle. Recommended for safe operation of HPLC systems.
Ordering information: Suitable for capillaries with 3.2 mm OD. For other diameters use other fittings.
Related Information
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
SCAT Europe Safety Cap Test Report.pdf
Product Literature
Customers Who Bought This Also Bought