SafetyCaps against Hazardous Vapours, S.C.A.T.
Supplier: Avantor
Certificates
About this item
Extract your solvents safely from bottles and reservoirs without being exposed to hazardous gases and solvent vapours.
- Especially recommended for HPLC Systems
- Chemically resistant against aggressive organic solvents
With integrated air valve for pressure equalisation. High quality materials (PTFE, PP) ensure maximum safety for your laboratory staff and environment.
Renew air valve twice a year for maximum safety.
SafetyCaps combined: The SafetyCap combi and its all-around connector provide a stable baseline and reliable reproducibility for RI detection. For fast and simple solvent degassing with helium.
Ordering information: All SafetyCaps are delivered with an air valve and corresponding fittings. For additional air valve options, see Air Valves with Filter Function for Safety Caps, S.C.A.T.
Related Information
GHS Poster Important Lab and HPLC Safety Guidelines
Application Notes
SCAT Europe Safety Cap Test Report.pdf
Product Literature
SCTV107910
Images 360
Customers Who Bought This Also Bought